ULTRA LC-MS SOLVENTS
New LC-MS Ultra Hipersolv CHROMANORM solvents that provides an outstanding quality for UHPLC-MS
LC-MS Ultra Hipersolv CHROMANORM grade solvents are exclusively tested for high performance with UHPLC gradient specifications plus UV, positive and negative mode MS detection.
- Tested specifically in UHPLC-MS with an extremelly small drift in UHPLC gradient analysis
- High purity for extremelly low detection limits under any detection mode together with UHPLC
- Positive and negative ion mode testing for all MS applications
- Low level of inorganic and metal ions for high sensitivity spectra
- Low gradient baseline and most narrow and consistent specifications
Log in to see your contract pricing and availability.