Melting point standards
To calibrate your equipment with accuracy
Product range is composed by 10 melting point standards from Benzophenone (melting Point 47 to 49 °C) to Anthraquinone (melting Point 283 to 286 °C)
  • The majority of the extensive range available in 1 g
  • Reliable, easy to use, accurate
  • Uncertainty of measurement up to ± 0.3 °C
  • Can be used with any melting point apparatus
  Description Pk Cat. No. Promo price Quantity
  Benzophenone, m.pt.= 47...49 °C 1 g 85101.001E  
  p-Nitrotoluene, m.pt.= 52...54 °C 1 g 85102.001E  
  Vanillin, m.pt.= 81...83 °C 1 g 85103.001E  
  Benzoic acid, m.pt.= 121...123 °C 1 g 85104.001E  
  Phenacetin, m.pt.= +133...135 °C 0.5 g 85105.005E  
  Salicylic acid, m.pt.= 158...160 °C 1 g 85106.001E  
  Sulphanilamide, m.pt.= 164 ...166 °C 1 g 85107.001E  
  Caffeine, m.pt.= 235...238 °C 1 g 85108.001E  
  Carbazole, m.pt.= 243...247 °C 1 g 85109.001E  
  Anthraquinone, m.pt.= 283...286 °C 1 g 85110.001E  
  Melting point standard set (Sulphanilamide, Caffeine, Vanillin) 3 g 85111.003E  
  Melting point standard set, Benzophenone (47...49 °C), Benzoic acid (121...123 °C), Anthraquinone (283...286 °C) 3 g 85112.003E  
  Melting point standard set, Vanillin (81...83 °C), Phenacetin (134...136 °C), Caffeine (235...237 °C) 3 g 85113.003E