Automate media transfer and save time: Masterflex pumps for cell culture efficiency
Boost the efficiency of your cell culture fluid handling with Masterflex pumps
Increase efficiency in your cell culture workflows when you use accurate, reliable Masterflex® pumps from Avantor®.
Reproducible, accurate media dispensing and sterile handling are critical to successful cell culture workflows. Masterflex pumps allow you to have more precise control over your volumes, save time with automation, and keep your cell culture medium contamination-free. Masterflex pumps are designed to streamline your workflow – learn how adding Masterflex pump solutions to your cell culture fluidics handling will enhance your cell culture operations today.
Why choose Masterflex pumps?
Time savings. All Masterflex pumps are programmable, so you can automate your repetitive fluid transfer tasks and reclaim valuable time.
Efficiency. With a user-friendly digital interface and plenty of memory for different fluid handling programs, Masterflex makes setting up repeatable processes easy.
Accuracy. See more reproducible volume dispensation for more accurate results with pumps that ensure less media waste and more consistent flow rates.
Robustness. Peristaltic pumps avoid introducing contamination and ensure low shear stress on your cells.
Cost savings. Spend less on supplies as better precision means less waste of buffers, serum, and media.
Masterflex offers versatile pumps for accurate low-flow fluid delivery. Peristaltic pumps reduce the shear force on your media, so your cells stay intact during transfer. Compact benchtop pumps optimize ease-of-use, accuracy, and reliability while the pump head accepts continuous tubing for clean, uninterrupted fluid paths for better sterile fluid handling.
For the most reproducible sterile cell culture fluid handling, build out your pump systems with high quality Masterflex tubing and pump heads. Whether scaling up experiments or maintaining consistent cell lines, Masterflex materials from Avantor provide the reliability and reproducibility you need.
Don't let slow, manual fluid handling hold you back. Transform your cell culture workflow with Masterflex and experience the benefits for yourself.
Upgrade your workflow
![](/m-uk.vwr.com/en_GB/images/Variable-Speed-Pump-200x200.png)
Masterflex® L/S® Variable Speed Pump with Easy-Load® Precision Pump Head
Looking for a straightforward solution for cell media handling?
The analogue L/S variable speed pump is a set-up-and-go solution for fluid handling for cell culture labs just starting to automate their processes. Compatible with Cytoflow® pump heads for reduced shear stress.
![](/m-uk.vwr.com/en_GB/images/Reglo-Digital-Miniflex-Pumps-200x200.png)
Masterflex® Ismatec® Reglo Digital Miniflex Pumps
Looking to reliably and reproducibly aliquot low volumes of media for smaller projects?
Miniflex pumps offer an easy-to-use digital touchscreen interface with programmable modes for more versatility, such as continuous or dispensing pumping and wide flow rate and dispense volume ranges.
![](/m-uk.vwr.com/en_GB/images/Reglo-Independent-Channel-200x200.png)
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps
Looking for a highly customizable cell culture fluid handling solution?
Reglo ICC pumps empower flexibility in your fluid handling capabilities, without increasing benchtop footprint. Get up to 4 flow channels, programmable with different rates, dispense volumes, and directions, to handle even the most complex cell culture experiments.
Try our Pump Selection Tool to guide you through selecting the right pumps, tubing, and connections for your needs
Masterflex for cell culture resources
How to choose the right Masterflex Pump system
The “STAMP” tubing selection guide
Shear-sensitive pumping with peristaltic pumps
Sterile, sterilization, and sterility – the language of keeping things contamination-free
Using masterflex L/S and I/P process pumps in ISO Class 5 (class 100) cleanroom applications