Melting point standards
To calibrate your equipment with accuracy
Product range is composed by 10 melting point standards from Benzophenone (melting Point 47 to 49 °C) to Anthraquinone (melting Point 283 to 286 °C)
- The majority of the extensive range available in 1 g
- Reliable, easy to use, accurate
- Uncertainty of measurement up to ± 0.3 °C
- Can be used with any melting point apparatus
Log in to see your contract pricing and availability.