Exciting changes are coming soon—your Punchout account will be redirected and migrated to a new platform on June 16 to enhance your experience! If your company requires our new web certificate and IP address, these can be found here at the very bottom of the page under ‘B2B Avantorsciences.com and Wardsci.com punchout IP and certificates’.
182 density standards for use at different temperatures (15°C, 20°C, 25°C, 40°C, 50°C, 60°, 80°C)
Extensive range of density standards in accordance with ASTM D1480-12 for density or relative density (specific gravity) measurements by Bingham Pycnometer.
High accuracy
Verified using Bingham Pycnometer
All products tested in accordance with ASTM D1480-12 guidelines
Consistent, traceable, certified
Certificates of Analysis and Safety Data Sheets available online
Log in to see your contract pricing and availability.