Cell culture incubators and water baths free of contamination
A5230 Incubator-Clean™ is a registered trademark of AppliChem GmbH.
  • Disinfectant solution for incubators and sterile benches
  • Fully compatible with common work surfaces
  • Non-toxic and biodegradable ingredients
Product Information
A5219 Incuwater-Clean™ is a registered trademark of AppliChem GmbH.
  • 100X concentrated solution (use 50 ml per 5 litres of incubator water bath)
  • Disinfectant solution for CO2 incubator water baths
  • For the prevention of microbial growth in incubator water baths
Product Information
A9390 Aquabator-Clean™ is a registered trademark of AppliChem GmbH.
  • 100X concentrated solution (use 10 ml per 1 litre of water)
  • Disinfectant solution for ordinary water baths (not for CO2 incubators)
  • For the prevention of microbial growth in water baths
Product Information
Request Quote