Sorbents for column chromatography, ISOLUTE® MSPD
Supplier: Argonaut Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9370-0100 9370-1000 9371-0100 9371-1000
ARGO9370-0100EA 0 GBP
ARGO9370-0100 ARGO9370-1000 ARGO9371-0100 ARGO9371-1000
Sorbents for column chromatography, ISOLUTE® MSPD
Chromatography Sorbents SPE Sorbents
Matrix solid phase dispersion is a sample preparation technique for use with solid sample matrices. Two MSPD grade ISOLUTE sorbents, MSPD C18 and MSPD C18(EC) have been developed by Argonaut and is particularly applicable to the extraction of basic veterinary drugs from liver and other biological matrices. These sorbents produce a more homogeneous and rapid blend of the matrix-sorbent mixture compared with SPE or preparative grade C18 sorbents.
  • Simple and quick for multiple extractions
  • Eliminates emulsion formation
  • Compatible with viscous matrices that can plug SPE columns