SPE columns, ISOLUTE® C18(EC)
Supplier: Biotage
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ISOLUTE®
ARGO221-0050-LEA 0 GBP
ARGO221-0050-L ARGO221-0020-A
SPE columns, ISOLUTE® C18(EC)
Chromatography Columns
ISOLUTE® C18(EC) is used for the extraction of acidic, neutral and basic compounds from aqueous sample matrices. It is a non-polar sorbent, manufactured using a trifunctional silane and endcapped to reduce the polar secondary interactions associated with surface silanol groups.