SPE columns, ISOLUTE® ENV+
Supplier: Biotage
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ISOLUTE®
ARGO915-0050-LEA 0 GBP
ARGO915-0050-L ARGO915-0010-B ARGO915-0020-CS ARGO915-0020-BS ARGO915-0001-G ARGO915-0020-H
SPE columns, ISOLUTE® ENV+
Chromatography Columns
ISOLUTE® ENV+ is a hyper crosslinked hydroxylated polystyrene-divinylbenzene copolymer, with very high surface area, ideal for extracting polar analytes from large volume water samples. The particle size and retentive nature of the sorbent allow exceptionally high sample flow rates to be used without reducing extraction efficiency or analyte recovery.
* Fitted with stainless steel frits