SPE columns, ISOLUTE® MFC18
Supplier: Argonaut Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
240-0020-B 240-0020-H 240-0500-E 9240-0100
ARGO240-0020-BEA 0 GBP
ARGO240-0020-B ARGO240-0020-H ARGO240-0500-E ARGO9240-0100
SPE columns, ISOLUTE® MFC18
Chromatography Columns
ISOLUTE® MFC18 is a retentive, non polar sorbent for extraction of a range of analytes from aqueous samples using a non polar retention mechanism. It is used for extraction from aqueous matrices using non polar interactions (for acidic, neutral and basic analytes) and readily accessible secondary silanol interactions (for basic compounds only). Not the ideal choice when working at extremes of pH, due to its relatively low carbon load.