Anti-CD36 Mouse Monoclonal Antibody [clone: FA6-152]

Supplier: STEMCELL Technologies
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

60084
STMC60084EA 198 GBP
STMC60084
Anti-CD36 Mouse Monoclonal Antibody [clone: FA6-152]
Antibodies
Mouse monoclonal IgG1 antibody against human, rat CD36, unconjugated.

  • Monocytes
  • Other

The FA6-152 antibody reacts with CD36, an ~88 kDa transmembrane glycoprotein that functions as a scavenger receptor, cell adhesion molecule, and signal transducer on the surface of many types of cells including platelets, monocytes, macrophages, erythrocyte precursors, endothelial and epithelial cells, and some macrophage-derived dendritic cells. The topology of the protein is thought to comprise a large extracellular loop with the N- and C-termini both insertedMore Product Information
Order Now

SPECIFICATIONS

Antigen name CD36
Environmentally Preferable
Clonality Monoclonal
Conjugation Unconjugated
Clone FA6-152
Reactivity Human,Rat
Host Mouse
Gene ID 948
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR