RoboSep™ Buffer

Supplier: STEMCELL Technologies
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

20104 20124
STMC20104EA 24 GBP
STMC20104 STMC20124
RoboSep™ Buffer
Cell Separation Media
RoboSep™ buffer is recommended for EasySep™ cell separation protocols performed by RoboSep™.

Application: Immunology
Species: Human, mouse, non human primate, other, rat

Please note that one or two bottles of buffer are included with every purchase of a RoboSep™ reagent kit.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission ofMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR