EasySep™ Mouse Mesenchymal Stem/Progenitor Cell Enrichment Kit
Supplier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
19771
STMC19771EA 786 GBP
STMC19771
EasySep™ Mouse Mesenchymal Stem/Progenitor Cell Enrichment Kit
Cell Separation Media
Immunomagnetic isolation of untouched mouse mesenchymal stem/progenitor cells
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Isolated cells are untouched
The EasySep™ Mouse Mesenchymal Stem/Progenitor Cell Enrichment Kit is designed to isolate mesenchymal stem/progenitor cells from mouse compact bone by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-mesenchymal stem/progenitor cells (CD45, TER119). labelled cells are then recognized by Tetrameric Antibody Complexes that areMore Product Information