EasySep™ Human T Cell Enrichment Kits
Supplier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
19051 19051RF
STMC19051EA 709 GBP
STMC19051 STMC19051RF
EasySep™ Human T Cell Enrichment Kits
Cell Separation Media
Immunomagnetic isolation of untouched human T cells.
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Untouched, viable cells
The EasySep™ Human T Cell Enrichment Kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-T cells and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a newMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable