EasySep™ Release Human APC Positive Selection Kit
Supplier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100-0031
STMC100-0031EA 700 GBP
STMC100-0031
EasySep™ Release Human APC Positive Selection Kit
Cell Separation Media
Immunomagnetic positive selection of particle-free human cells labeled with APC-conjugated antibodies.
  • Highly purified cells labelled with APC-conjugated antibodies isolated from human samples in less than 40 minutes
  • No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Release Human APC Positive Selection Kit is designed to isolate cells that are labeled with APC-conjugated antibodies from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection.
Desired cellsMore Product Information