NeuroCult™ NS-A Differentiation Kit (Human)
Supplier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05752
STMC05752EA 236 GBP
STMC05752
NeuroCult™ NS-A Differentiation Kit (Human)
Cell Culture Media
Medium for differentiation of human neural stem and progenitor cells.
NeuroCult™ NS-A differentiation kit (Human) is a standardised medium for the differentiation of human neural stem and progenitor cells into neurons, astrocytes, and oligodendrocytes.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').