Anti-UPP2 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

55-545
PRSI55-545EA 604 GBP
PRSI55-545
Anti-UPP2 Rabbit Polyclonal Antibody
Antibodies
Catalyzes the reversible phosphorylytic cleavage of uridine and deoxyuridine to uracil and ribose-or deoxyribose-1-phosphate. The produced molecules are then utilized as carbon and energy sources or in the rescue of pyrimidine bases for nucleotide synthesis. Shows substrate specificity and accept uridine, deoxyuridine, and thymidine as well as the two pyrimidine nucleoside analogs 5-fluorouridine and 5-fluoro-2(')-deoxyuridine as substrates.

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: UPP2
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR