Anti-GPX3 Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
29-588
PRSI29-588EA 643 GBP
PRSI29-588
Anti-GPX3 Rabbit Polyclonal Antibody
Antibodies
GPX3 belongs to the glutathione peroxidase family, which functions in the detoxification of hydrogen peroxide. It contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon, which normally signals translation termination.Glutathione peroxidase catalyzes the reduction of hydrogen peroxide, organic hydroperoxide, and lipid peroxides by reduced glutathione and functions in the protection of cells against oxidative damage. Human plasma glutathione peroxidase has been shown to be a selenium-containing enzyme. GPX3 expression appears to be tissue-More Product Information