Anti-PF-04518600 Recombinant Antibody

Supplier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10-982
PRSI10-982EA 637 GBP
PRSI10-982
Anti-PF-04518600 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: VH-CH1-VH-V-KAPPA-CH2-CH3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR