Masterflex® L/S® Analog Variable-Speed Console Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX77910-45-EUEA 2080 GBP
MFLX77910-45-EU MFLX77910-55-EU MFLX77910-65-EU MFLX77910-75-EU MFLX77910-45-UK MFLX77910-55-UK MFLX77910-65-UK MFLX77910-75-UK
Masterflex® L/S® Analog Variable-Speed Console Drive Systems, Avantor®
Pumps Peristaltic Pumps
Compact, cost effective pump systems for general fluid transfer applications.
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
Choose a convenient, complete system all with one catalog number.