Masterflex® L/S® Analog Variable-Speed Console Drive with Easy-Load® II Pump Head for Precision Tubing; 230 VAC
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX77910-45-EU
Your Organization's Price:
Retrieving
Compact, cost effective pump systems for general fluid transfer applications.
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
  • Flow rate 56 to 1700 mL/min with included L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
Choose a convenient, complete system all with one catalog number.
<b>Delivery information: </b>Includes console drive 07555-05, Easy-Load® II pump head 77200-62, 10-ft C-Flex® ULTRA® tubing, and 6-ft (1.8 m) line cord with IEC 320/CEE22 socket. Shipped with country-specific cord/plug set; please specify ultimate destination when ordering.