Flowline Plastic Mini-Float Level Switches
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
43299-24 43299-26
MFLX43299-24EA 34.7 GBP
MFLX43299-24 MFLX43299-26
Flowline Plastic Mini-Float Level Switches
Level Sensors
Control liquid level even in corrosive environments.
  • Excellent for clean, non-coating or non-scaling liquids
Choose either vertical or horizontal mounting to fit your application. Reliable reed switch output for interface with PLC or relay control devices. Your liquid must have a specific gravity of 0.8 or greater.
Delivery information: Includes submersible float with 2-ft (0.6-m) cable.