Flowline LV20-1201 Mini-Float Level Switch, Polypropylene, 1/8" NPT(M)
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX43299-24
Your Organization's Price:
Retrieving
Control liquid level even in corrosive environments.
  • Excellent for clean, non-coating or non-scaling liquids
Choose either vertical or horizontal mounting to fit your application. Reliable reed switch output for interface with PLC or relay control devices. Your liquid must have a specific gravity of 0.8 or greater.
<b>Delivery information: </b>Includes submersible float with 2-ft (0.6-m) cable.