Miniature inert valves, HV series
Supplier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
86725 86726 86728 86729 86730 86731 86732 86735 86736 86738 86788 86779 86777 86901 86903 86915 86778 86781 86775 86780 86918 86785 86786 86787 86902 86905 86919 86907 86910 86911 86912 86913 86916 86904
HAMI86725EA 138 GBP
HAMI86725 HAMI86726 HAMI86728 HAMI86729 HAMI86730 HAMI86731 HAMI86732 HAMI86735 HAMI86736 HAMI86738 HAMI86788 HAMI86779 HAMI86777 HAMI86901 HAMI86903 HAMI86915 613-1617 HAMI86781 HAMI86775 HAMI86780 HAMI86918 HAMI86785 HAMI86786 HAMI86787 HAMI86902 HAMI86905 HAMI86919 HAMI86907 HAMI86910 HAMI86911 HAMI86912 HAMI86913 HAMI86916 HAMI86904
Miniature inert valves, HV series
Valves Micro Valves
HV plug valves provide an economical and flexible method of custom-building a fluid flow control system, from a simple on/off valve to a more complex multi-valve arrangement, using a combination of loop and distribution valves.
  • Wetted paths are chemically inert PTFE and Kel-F® materials
  • Valve position is controlled manually via a handle attached to the valve stem
  • Valve can be chosen by selecting the proper flow path
The small valve body with its chemically inert flow path is ideally suited for low pressure applications. All HV Plug Valves allow you toMore Product Information