Removable needles
Supplier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
200285 200295 202645 207434 207419 7768-01 7780-01 7784-03 7780-04 7758-03 7770-01 7758-02 7784-06 7758-04 7780-03 7786-01 7784-01 7784-05 7779-01 7803-05 7806-01 7803-07 7758-01 7731-03 7779-03 7779-02 7804-02 7804-01 7762-06 7806-02 7804-04 7803-04 7770-02 7803-03 7768-02 7804-03 7784-02 7784-04 7731-01 7731-02 7731-04 7732-01 7803-02 7806-03 7762-04 7784-07 203481 8649-01 8650-01 7784-08 7786-02 7787-01 7787-02 7762-05 7780-02 7779-04 7762-03 7806-04 7732-02 7732-03 7732-04 7803-01 7806-06 7806-07 7804-10 7804-11 7806-10 7731-11 7803-06 7804-08 7806-11 7804-05 7804-06 7804-07 7804-09 7804-12 7806-05 7806-08 7806-09 209221 207434-10
HAMI200285EA 111 GBP
HAMI200285 HAMI200295 549-1059 HAMI207434 HAMI207419 HAMI7768-01 HAMI7780-01 HAMI7784-03 HAMI7780-04 113-0699 548-1563 113-0698 HAMI7784-06 HAMI7758-04 HAMI7780-03 549-0578 HAMI7784-01 HAMI7784-05 HAMI7779-01 HAMI7803-05 HAMI7806-01 HAMI7803-07 HAMI7758-01 HAMI7731-03 HAMI7779-03 HAMI7779-02 HAMI7804-02 HAMI7804-01 HAMI7762-06 HAMI7806-02 HAMI7804-04 HAMI7803-04 HAMI7770-02 HAMI7803-03 HAMI7768-02 HAMI7804-03 HAMI7784-02 HAMI7784-04 HAMI7731-01 HAMI7731-02 HAMI7731-04 HAMI7732-01 HAMI7803-02 HAMI7806-03 HAMI7762-04 HAMI7784-07 HAMI203481 HAMI8649-01 HAMI8650-01 HAMI7784-08 549-0579 HAMI7787-01 HAMI7787-02 HAMI7762-05 HAMI7780-02 HAMI7779-04 HAMI7762-03 HAMI7806-04 HAMI7732-02 HAMI7732-03 HAMI7732-04 HAMI7803-01 HAMI7806-06 HAMI7806-07 HAMI7804-10 HAMI7804-11 HAMI7806-10 HAMI7731-11 HAMI7803-06 HAMI7804-08 HAMI7806-11 HAMI7804-05 HAMI7804-06 HAMI7804-07 HAMI7804-09 HAMI7804-12 HAMI7806-05 HAMI7806-08 HAMI7806-09 HAMI209221 HAMI207434-10
Removable needles
Syringe Needles
The benefit of a replaceable needle for the occasional bend or clog and the minimal dead volume of a cemented needle. The unique removable needle design installs the needle precisely at the zero line. The result is a zero dead volume connection that simplifies priming.
Custom lengths and point styles are available. You can order your custom made needle according to your specification.
* Large hub
** Kel-F hub