NHS Ester, SureLight® stain_Molecular Biology

Supplier: Columbia Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D44-010-100UG
COBSD44-010-100UGEA 46.3 GBP
COBSD44-010-100UG
NHS Ester, SureLight® stain_Molecular Biology
NHS Ester

SureLight® 488 NHS Ester is a bright green fluorescent dye with excitation suited to the 488nm laser line, fluorescent microscopy and flow cytometry. It offers a much more stable signal generation in imaging and flow cytometry than FITC and is half the cost of Alexa Fluor® 488 with no royalties or licensing fees.


  • Excitation max 486 nm
  • Emission max 517 nm


The NHS-ester version is an efficient amine reactive probe that can be used to label proteins, antibodies, haptens and other primary amine containing molecules.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR