Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: Columbia Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D3-113-FAB
COBSD3-113-FABEA 375 GBP
COBSD3-113-FAB
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation APC (Allophycocyanin)
Reactivity Rat
Host Goat
Isotype IgG
ELISA Yes
Flow cytometry Yes
Cross adsorption Bovine, Rabbit
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR