MagaDye™ 561 ddATP
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17062 17066
AATB17062EA 1250 GBP
AATB17062 AATB17066
MagaDye™ 561 ddATP
Nucleic Acid Reagents Sanger Sequencing Reagents
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Caution: For research use only.