Glutamic-C™, sequencing grade
Supplier: G Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-15
786-15EA 590 GBP
786-15
Glutamic-C™, sequencing grade
Nucleic Acid Reagents Sanger Sequencing Reagents
SG-Glutamic-C™ is a serine endopeptidase, from S. aureus V8, that is highly specific for the cleavage of peptide bonds at the carboxy side of either aspartic or glutamic acid, depending on the buffer used. In Tris-HCl buffer, in particular in the absence of phosphate ions, the enzyme is specific for the glutamyl site. Recommended buffers for fragmentation of proteins using this enzyme are 50 mM Tris-HCI, pH 8,0 or bicarbonate buffer. Highly purified preparations of SG-Glutamic-C™ are chemically modified making the enzyme both resistant to autolysis and stabilises its enzymatic activity.
  • Modified glutamic-C for sequence analysis
  • Resistant to autolysis and degradation
  • High specificity, free from other known endopeptidases
  • Consistency of activity levels from lot to lot for reproducible digestions
SG-Glutamic-C™ is supplied lyophilised in 10 μg vials. The enzyme is typically reconstituted to a concentration of 0,5 μg/ml and commonly used at a ratio of 1:100 to 1:20 (enzyme to protein, by weight) in a standard digestion buffer.