Microlab 600 Bubble Free Prime (BFP) syringes
Supplier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
59000-05 59000-10 59000-15 59000-20 59000-25 59000-30 59000-35 59000-40 59000-45 59000-50 59000-55 59000-60 208336 208335 208337
613-1570EA 239 GBP
613-1570 613-1571 613-1572 613-1573 613-1574 613-1575 613-1576 613-1577 613-1578 613-1579 613-1580 613-1581 HAMI208336 HAMI208335 HAMI208337
Microlab 600 Bubble Free Prime (BFP) syringes
Syringes Syringes for Chromatography
The Bubble Free Prime (BFP) syringe is specifically designed for the Microlab® 600 series and the only syringe type compatible with the instrument. It has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
SaltLine syringes should be used when working with solutions that have a high salt concentration.
Order Now
SPECIFICATIONS
Environmentally Preferable