Headspace syringes for PAL COMBI–xt® headspace autosamplers
Supplier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
203082 203084 203086 203141 203181 203182
549-1324EA 225 GBP
549-1324 549-1325 549-1326 549-1327 549-1328 549-1329
Headspace syringes for PAL COMBI–xt® headspace autosamplers
Syringes Syringes for Chromatography
These HD headspace syringes feature a unique plunger designed specifically for the CTC PAL System COMBI xt® gas chromatography (GC) autosampler system. The High Dynamic (HD) plunger is optimised for high throughput and sets a new standard for headspace syringes.
  • Unique spring-in-plunger design creates a dynamic seal between the plunger tip and the inside of the glass barrel for leak-free operation
  • Excellent performance over a large temperature range
  • Increased lifetime compared to traditional headspace syringes increases value
  • Improved accuracy and
More Product Information