VWR Products
VWR circulating chillers
These circulating chillers are an environmentally-friendly, water-conserving and reliable means of controlling the temperature of liquids in open tanks or closed loop systems.
VWR refrigerated circulating baths with MX temperature controller
Stainless steel baths with user friendly MX temperature controller. Can also be used for external closed loop circulation. Cool Command™ technology regulates the amount of cooling required - saving energy.
VWR heating circulators
Stainless steel circulating baths with Advanced Programmable or Advanced Digital temperature controller. Variable speed pump with external circulation and temperature control capability.
VWR refrigerated/heated circulators
Stainless steel circulating baths with choice of Advanced Programmable or Advanced Digital temperature controller. Variable speed pump with external circulation and temperature control capability.