PESTINORM SUPRATRACE solvents for pesticide and organic substances trace analysis
PESTINORM SUPRATRACE – a universal solvent for every application
11 High purity solvents for all organic samples analysis and sensitive detection methods (GC-FID, GC-ECD, GC-MS)
- Very high purity (>99.9% of assay), Reliability
- One universal solvent for all samples and sensitive detection methods
- Specified retention time range for ECD is larger (so even low-boiling substances can be reliably detected)
- All detection methods included in specifications
Log in to see your contract pricing and availability.