Disposable PPE garments for use in your cleanroom - sterile and non sterile
Available as individual coveralls or kits (sterile)
Protecting the operator as well as product and process in your cleanroom.
- Coveralls (microporous) with hood Cat III type 4/5/6 (IEST -RP-CC003,3 - Level II)
- Coveralls (SMS) with hood Cat III type 5/6, sterile and non sterile (IEST -RP-CC003,3 - Level I)
- Also available as a sterile kit including coverall, hood, boots and sleeves
- Each piece individually double packed
Log in to see your contract pricing and availability.