Software & Adapters
All USB and HDMI/USB cameras require a software which can be downloaded from the product pages.
HDMI cameras offer settings directly from the device by using a mouse when being used without a PC.
Tablet cameras have a Tablet App so do not need an extra software to be downloaded.
C-mount adapters
C-mount adapters exploit the physical connection between trinocular tube of the microscope and the digital camera.
Adapters have different magnification capability. It is important to choose the correct adapter magnification which depends on camera sensor size.
Download the selector guide to find the right connection between the VWR®camera and the VisiScope® microscopes.
Software versions
VisiCam® Vision Lite Soft is the most intuitive software for basic needs: capture images & stream live videos, perform linear measurements, export comprehensive reports.
VisiCam® Proview Soft which is a professional image analysis with a lot of additional parameters.
VisiCam® image analysis software: Intuitive software for professional image analysis.
Find here the overview of the software features and a selector to use the correct software for the correct camera.
The software can be downloaded for free on the camera product pages.
Cameras and Software
Camera | Visicam Vision Lite (free download) |
Visicam Vision Proview (free download) |
Visicam Image Analysis (free download) |
Camera VisiCam® B5 | X | ||
Camera VisiCam® 3 + | X | ||
Camera VisiCam® 5 + | X | ||
Camera VisiCam® 16 + | X | ||
Camera VisiCam® 20 + | X | ||
Camera VisiCam® 6 + | X | ||
Camera VisiCam® P6 | X | X | |
X | |||
Camera VisiCam® MET 5000 | X | ||
Camera VisiCam® HRS 14 | X | ||
Camera HDMI / USB VisiCam® HDMI5 | X | ||
Camera HDMI / USB VisiCam® HDMI6 | X | ||
Camera HDMI / USB VisiCam® HDMI HP | X | X | |
Wifi / USB camera VisiCam® 83 | X | ||
Wireless Tablet camera VisiCam® TC 20 + | Software tablet application | ||
Tablet TC 210 | X | X |
Software and image analysis
Features | Vision Lite |
Proview | Image Analysis |
Friendly Interface, multilanguage | Yes, 8 languages | Yes, 8 languages | Yes |
Capture still images and Stream live videos | Yes | Yes | Yes |
Images saved in different formats | Yes | Yes | Yes |
Image stack acquisation | Yes | Yes | Yes |
Grid addition | Yes | Yes | Yes |
Perform linear measurements ( line, circle, rectangle, polygon, angles, …) | Yes | Yes | Yes |
Accurate measurements through simple calibration | Yes | Yes | Yes |
Save Measurement results | Yes | Yes | |
Comprehensive data export ( notes and measures included ) | Yes | Yes | Yes |
Export comprehensive reports | Yes | Yes | Yes |
White & Black balanceo | No | Yes | Yes |
Exposure time | No | Yes | Yes |
Colors channels adjustment, contrast, saturation, brightness | No | Yes | Yes |
Management of several cameras | No | Yes | Yes |
Fully customizable graphical user interface ( password protection .. ) | No | Yes | No |
Fluorescence imaging with "pixel shift " function | No | Yes | Yes |
High Dynamic range ( HDR) acquisition | No | Yes | Yes |
Extended depth of focus ( EDF) | No | Yes | Yes |
Image stitching | Yes | Yes | |
Compare live and saved image | Yes | ||
Time Lapse capture | Yes | ||
Counting | Yes |