Isolator Cleaning
Cleaning of Compounding Aseptic Isolators (CAIS) and ClassII Biological Safety Cabinets (BSCS)
Compounding Aseptic Isolators and Class II biological safety cabinets are used in hospital pharmacies and other dispensing facilities to formulate (i.e. “compound”) individual prescriptions for patient care. A typical hospital pharmacy utilizing a CAI for contamination control and sterility, will make individual pharmaceutical preparations that are transferred aseptically into vials, intravenous (IV) bags, syringes, infusion devices, etc.