Leica Advanced Microscopy
Sample Preparation Through Publication Quality Imaging
Research workflows from Leica Microsystems help you save time and drive new discoveries. From automated sample preparation systems through to advanced imaging solutions, Leica products help you reduce costs and save time in drug discovery, tissue culture, live cell and translational research. Make your next breakthrough with innovative products from Leica Microsystems.
Leica Mica, the world’s first imaging Microhub
Mica unites widefield and confocal imaging in a sample protecting, incubating environment. With the simple push of a button, you have everything you need - all in one place - to supercharge fluorescence imaging workflows and get meaningful scientific results faster.
Leica THUNDER Imager, Organism
The THUNDER Imager Organism model allows fast and easy 3D exploration of whole organisms for developmental or molecular biology research.
Leica THUNDER Imager, Tissue
The THUNDER Imager Tissue model allows real time fluorescence imaging of 2D tissue sections typically used in neuroscience and histology research. Acquire rich, detailed images of thick tissues free of haze from 'out of focus' blur.
Leica THUNDER Imager, 3D Assay
The THUNDER Imager 3D Assay provides you with a solution for advanced 3D cell culture assays, whether you want to study stem cells, spheroids or organoids.
Leica Laser Induced Breakdown Spectroscopy (LIBS)
The Leica DM6 M LIBS offers two systems in one for visual and chemical analysis. It takes 1 second to have a chemical fingerprint withouth sample preparation. The integrated laser spectroscopy function delivers the chemical composition of the microstructure that you see in the microscope image.
Upright Materials Microscopes Leica DM4 M & DM6 M
Do you need to image, measure, and analyze similar features across many samples and materials science and analysis? The Leica DM4 M and Leica DM6 M are the Industrial Microscopes for you, whether microscopy novice or professional.
*Use the Leica DM4 M for manual routine inspection
*Use the Leica DM6 M for fully automated materials analysis
You can easily recall previous microscope settings and instantly reproduce imaging parameters with the unique Store and Recall software functions. For every sample type.
One-button Intelligent Automation makes your repetitive work easy. Reduce training time, improve your workflow, and get brilliant imaging results every time with the microscopes that don’t forget.
Leica Laser Microdissection (LMD)
Laser Microdissection (LMD) enables users to isolate specific single cells or entire areas of tissue. Powered by a unique laser design and dynamic software, Leica LMD systems allow users to easily isolate Regions Of Interest (ROI) from entire areas of tissue down to single cells or even subcellular structures such as chromosomes.
DMi8 - Modular inverted microscope for widefield research
Whether you need to precisely follow the development of a single cell in a dish, screen through multiple assays, obtain single molecule resolution, or tease out behaviors of complex processes, a DMi8 S system enables researchers to see more, see faster, and find the hidden.
*See more – Increase your viewing area up to 10,000x
*See faster – Experiments up to 5x faster
*See the hidden – Activate, ablate, and bleach within one experiment
Leica Cell DIVE multiplexed imaging solution
Enable new discoveries that improve patient outcomes through a deeper understanding of tissue microenvironments. Spatially map 60+ biomarkers from just one tissue section, at the single-cell level.