Lauda products
Lauda circulating chillers, Microcool
Range of circulation chillers includes five compact models providing cooling capacities from 0,25 to 1,2 kW for external applications. The units are ideal for use where heat needs to be dissipated reliably and quickly.
Lauda heating thermostats with transparent baths, ECO
ECO units with transparent plastic baths provide visibility where test samples need to be observed. Flow rate switch for flow rate control between internal and external circulation.
Lauda cooling/heating circulation thermostats, thermoelectric, LOOP
The LOOP product range includes two thermo-electric circulation thermostats for external temperature control. Refrigerant-free temperature control with Peltier technology.