Samplers For Viscous Materials
Samplers for medium viscous substances like oils, emulsions, creams, slurries and semi-solid materials, such as cheese, butter, clay, soap etc.
![](/m-uk.vwr.com/en_GB/images/viscous_medias_samplers_140.jpg)
Samplers for viscous medias - syringe principle
Strong suction power for viscous media. The media is drawn up and discharged via the tip of the sampler for easy sampling.
![](/m-uk.vwr.com/en_GB/images/spatulas_140.jpg)
Spatulas
Tools for taking samples of pasty materials for collecting, dividing and mixing pastes, creams and other viscous materials.
![](/m-uk.vwr.com/en_GB/images/lances_viscous_materials_140.jpg)
Lances and hand drills for viscous materials
Ideal for cutting out samples for quality control from paste-like substances. Available in different versions for light- or heavy-sticking products.