Sampler For Bulk Goods
Tools for sampling of bulk materials. Find sampling solutions for finest powders, granules, or large grains such as corn or nuts.
Samplers for bulk goods - sampling lances with inner and outer tubes
Wide range of sampling lances for point, multi-point or cross-sectional samples of any kind of bulk goods. Made from high quality materials.
- Zone samplers - selection of samplers with open inner tubes and closed chambers
- Spiralus - allows stepwise opening of the sampling lance
- EasySampler - easy to empty
- Scratcher - for small amounts of non pourable substances
- All-layer Scratcher - for non pourable substances
- All-layer Drill - for heavily compacted materials
Bag / sack samplers
Ideal for taking samples directly out of packaging and collecting them in their corresponding lances, sampling bottles or bags.
Samplers for silos and silo trucks
To take a representative sample out of a silo or silo truck, samplers often need to be extendable and very robust. See our selection here.
PowderProof - electric sampling device
This electric sampler is a convenient solution for taking vertical and horizontal samples of powders and granules from drums, bags or sacks...
Samplers for very small sampling amounts
Very expensive materials or ingredients that demand that the sample taken is very small but still representative, with minimum loss of material.