Alcohols, Disinfectants and Detergents
Contec offers a wide range of specially formulated chemicals for the cleaning and disinfection of critical and sterile environments. From hydrogen peroxide solutions and IPA, to fast-acting validated sporicidal and broad-spectrum disinfectants, Contec has the cleaning solutions you need. Contec can also offer a sterile neutral detergent solution ideal for cleanroom environments.
Featured products
Contec ProChlor Sporicidal Disinfectant
Contec ProChlor is a revolutionary sporicide achieving a 100% kill against spores in under 1 min. A blend of hypochlorous acid in purified water, Contec ProChlor is provided ready-to-use and is efficacious against bacteria, fungi, viruses and spores.
Contec HydroPure No Residue Disinfectant
Contec HydroPure is a blend of 6% H2O2 and WFI or purified water. Efficacious against bacteria, fungi and spores, Contec HydroPure leaves little to no residue. Available sterile with a guaranteed endotoxin level making it ideal for product contact areas.
Contec Sterile 70% Alcohol Range
Contec 70% IPA or denatured ethanol is available filtered and sterile in either 500ml or 1L trigger sprays, or 5L capped containers. Cleanroom processed, filtered to 0.2 micron and either double or tripled wrapped.
Contec NeutraKlean Detergent
Disinfection is more effective on a pre-cleaned surface. Contec NeutraKlean is a gentle and non-foaming detergent designed for cleanrooms. It is ideal for the low level cleaning required after maintenance and to remove disinfectant residues.