Certified membranes for microbiological analysis
![]() |
Membrane refills for Sentino® membrane dispenser Sized to minimise storage space resulting in less packaging material waste. Mixed cellulose esters is the most accepted filter media for microbiological analysis, and provides maximum recovery of organisms. High Flow Rate membrane optimised for biological, pharmaceutical, and sterilising filtration requirements. High Flow Rate membrane with dark background provides excellent contrast for counting opaque colonies. Exclusive dot grid pattern does not enhance or inhibit colony growth.
|