The ultimate combination of speed, sensitivity and flexibility
Applications
- High content toxicity assays
- Imaging spheroids and other 3D cellular models
- Characterisation of 3D neuronal networks
- Zebrafish assays
- Characterisation of sub-cellular processes and structures
ImageXpress® Micro 4 and ImageXpress® Micro Confocal
The ImageXpress Micro 4 is a versatile, customizable widefield instrument, equipped with a large field of view camera and an on-demand solid state light engine, providing exceptionally high throughput. The ImageXpress Micro 4 system offers unlimited configurability with user exchangeable filter cubes, the widest range of objective lenses and environmental controls, transmitted light and fluidics options. It also offers a field upgradeable option to add confocal imaging capability.
Complex, three-dimensional cellular models yield more predictive, physiologically relevant results versus monocultures or other two-dimensional cellular models. Explore the complexities of these models faster and gain better results even in samples grown in a thick extracellular matrices using the ImageXpress Micro Confocal.
Supported by highly customizable and powerful MetaXpress® High-Content Image Acquisition and Analysis Software, the ImageXpress systems provide a fast and robust platform to deliver seamless workflows from image acquisition to data analysis and to translate new discoveries into scientific breakthroughs.