Microflex® 93-260 nitrile and neoprene double-coated disposable gloves
Thinnest chemical resistant synthetic composite disposable glove
- Three layer design for superior protection against harsh chemicals including acids, solvents and bases
- Thin mil construction provides enhanced tactility and dexterity
- Extra soft material and ergonomic design for outstanding fit, feel and flexibility for longer wear time
- Lower acceptable pin hole rate (0,65 AQL) and extended cuff for reliable protection against hazardous substances
- Silicone-free formulation and processing ensure better product protection