Bodyguards® GL890 Examination Gloves, Nitrile, Powder Free
Supplier: Local
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GL8903B GL8904B GL8905B GL8902B GL8901B
LOCAGL8903BEA 45.6 GBP
LOCAGL8903B LOCAGL8904B LOCAGL8905B LOCAGL8902B LOCAGL8901B
Bodyguards® GL890 Examination Gloves, Nitrile, Powder Free
Gloves
The GL890 Bodyguards® Blue Nitrile Gloves are mid-weight, non-sterile medical gloves designed for reliable performance and user comfort. Featuring optimised textured finger grip, these gloves ensure precise and confident handling of instruments and tools, enhancing both control and tactile sensitivity. Made from a soft elastic material, they provide superior flexibility and comfort, while being latex free to suit individuals with latex allergies.
  • Latex free: No latex proteins - eliminates protein sensitisation
  • Textured finish to the fingers to enhance grip and handling
  • Nitrile technology provides an exceptionally soft and pliable material
  • Latex free, reducing the risk of hypersensitivity and allergic reactions to natural rubber proteins
  • powder free, reducing the risk of dust contamination
Certifications:
AQL 1.5. Compliant with European Standards EN455: Medical Gloves for Single Use parts 1,2,3&4. Compliant with Medical Devices Regulation (EU) 2017/745 Class 1. EU Certified. Manufactured in accordance with Personal Protective Equipment (PPE) by the European PPE Regulation (EU) 2016/425. Tested in compliance with Commission Regulation (EU) No 10/2011 for Food contact. Tested to European standards EN ISO 21420. EN ISO 374-1. EN ISO 374-4 & EN ISO 374-5.
Packaging:
Packing contents - All: 100 gloves per dispenser, 10 dispensers per case, 50 cases per pallet.