Anti-Pyruvate Dehydrogenase E2 Rabbit Monoclonal Antibody [clone: EPR12455(B)]

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB172617-100 AB172617-1ML
ABCAAB172617-100EA 458 GBP
ABCAAB172617-100 ABCAAB172617-1ML
Anti-Pyruvate Dehydrogenase E2 Rabbit Monoclonal Antibody [clone: EPR12455(B)]
Antibodies
Rabbit monoclonal [EPR12455(B)] to Pyruvate Dehydrogenase E2.

Type: Primary
Antigen: Pyruvate Dehydrogenase E2
Clonality: Monoclonal
Clone: EPR12455(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR