Anti-IFN gamma Receptor beta/AF-1 Rabbit Monoclonal Antibody [clone: EPR22171]

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB224197-100 AB224197-1ML
ABCAAB224197-100EA 489 GBP
ABCAAB224197-100 ABCAAB224197-1ML
Anti-IFN gamma Receptor beta/AF-1 Rabbit Monoclonal Antibody [clone: EPR22171]
Antibodies
Rabbit monoclonal [EPR22171] to IFN gamma Receptor beta/AF-1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: IFN gamma Receptor beta/AF-1
Clonality: Monoclonal
Clone: EPR22171
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Recombinant full length protein
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR