Chromone-2-carboxylic acid ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Oxo-4H-1-benzopyran-2-carboxylic acid, Chromocarb, 4-Oxo-4H-chromene-2-carboxylic acid
A13552.06
A13552.14
A13552.06EA
17.7
GBP
A13552.06
A13552.14
Chromone-2-carboxylic acid ≥97%
Chromone-2-carboxylic Acid
Purity: 97%
CAS No.: 4940-39-0
Molecular Formula: C10H6O4
Formula Weight: 190.15
Melting Point: ca 260° dec.
Beilstein Reference: 146442
Merck Reference: 14,2237
MDL No.: MFCD00006838
Formula:
C₁₀H₆O₄ MW: 190.15 g/mol Melting Pt: ∼ 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006838 CAS Number: 4940-39-0 EINECS: 225-583-0 Merck Index: 14,2237 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...