4-(Trifluoromethylthio)phenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008719-1G 008719-5G
FLUO008719-1GEA 0 GBP
FLUO008719-1G FLUO008719-5G
4-(Trifluoromethylthio)phenol
4-(Trifluoromethylthio)phenol
Formula: C₇H₅F₃OS
MDL Number: MFCD00036035
CAS Number: 461-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR