Bis(tri-tert-butylphosphine)palladium(0) ≥98%
Supplier: abcr
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Bis(tri-tert-butylphosphine)palladium, Bis(tri-t-butylphosphine)palladium(0)
AB134604
S46-0252.0250
AB134604.0001
ABCRAB134604EA
0
GBP
ABCRAB134604
ABCRS46-0252.0250
ABCRAB134604.0001
Bis(tri-tert-butylphosphine)palladium(0) ≥98%
Bis(tri-tert-butylphosphine)palladium(0)
Formula:
C₂₄H₅₄P₂Pd |
MDL Number:
MFCD03094580 CAS Number: 53199-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...